4,5-Dibrom-2-fluortoluol

Lieferant: CHEMPUR

Synonyme: 1,2-Dibromo-4-fluoro-5-methylbenzene

FL13296.POR
CHMPFL13296.POREA 0 EUR
CHMPFL13296.POR
4,5-Dibrom-2-fluortoluol
4,5-Dibrom-2-fluortoluol
Formel: C₇H₅Br₂F
Molecular Weight: 267,92 g/mol
MDL: MFCD00143234
CAS-Nummer: 202982-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR