2,6-Dichlor-4-fluoranilin

Lieferant: CHEMPUR

FL13298.POR
CHMPFL13298.POREA 0 EUR
CHMPFL13298.POR
2,6-Dichlor-4-fluoranilin
2,6-Dichlor-4-fluoranilin
Formel: C₆H₄Cl₂FN
Molecular Weight: 180,01 g/mol
MDL: MFCD00142845
CAS-Nummer: 344-19-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR