3',4'-Difluoracetanilid

Lieferant: CHEMPUR

Synonyme: 3,4-Difluoroacetanilide, N-(3,4-Difluorophenyl)acetamide

FL13299.POR
CHMPFL13299.POREA 0 EUR
CHMPFL13299.POR
3',4'-Difluoracetanilid
3',4'-Difluoracetanilid
Formel: C₈H₇F₂NO
Molecular Weight: 171,1441 g/mol
MDL: MFCD00017903
CAS-Nummer: 458-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR