1,2-Difluor-4,5-dimethoxybenzol

Lieferant: CHEMPUR

FL13301.POR
CHMPFL13301.POREA 0 EUR
CHMPFL13301.POR
1,2-Difluor-4,5-dimethoxybenzol
1,2-Difluoro-4,5-dimethoxybenzene
Formel: C₈H₈F₂O₂
Molecular Weight: 174,15 g/mol
MDL: MFCD00070789
CAS-Nummer: 203059-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR