3,4-Difluorphenetol

Lieferant: CHEMPUR

Synonyme: 1,2-Difluor-4-ethoxybenzol, (3,4-Difluorphenyl)ethylether

FL13302.POR
CHMPFL13302.POREA 0 EUR
CHMPFL13302.POR
3,4-Difluorphenetol
3,4-Difluorphenetol
Formel: C₈H₈F₂O
Molecular Weight: 158,15 g/mol
MDL: MFCD00673005
CAS-Nummer: 163848-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR