4,5-Dimethoxy-2-fluorbenzonitril

Lieferant: CHEMPUR

FL13308.POR
CHMPFL13308.POREA 0 EUR
CHMPFL13308.POR
4,5-Dimethoxy-2-fluorbenzonitril
4,5-Dimethoxy-2-fluorbenzonitril
Formel: C₉H₈FNO₂
MDL: MFCD00143236
CAS-Nummer: 119396-88-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR