2-Fluorbenzaldehydoxim

Lieferant: CHEMPUR

FL13313.POR
CHMPFL13313.POREA 0 EUR
CHMPFL13313.POR
2-Fluorbenzaldehydoxim
2-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
Molecular Weight: 139,13 g/mol
MDL: MFCD00019958
CAS-Nummer: 451-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR