3-Fluorbenzaldehydoxim

Lieferant: CHEMPUR

FL13314.POR
CHMPFL13314.POREA 0 EUR
CHMPFL13314.POR
3-Fluorbenzaldehydoxim
3-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
CAS-Nummer: 458-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR