4-Fluorbenzaldehydoxim

Lieferant: CHEMPUR

FL13315.POR
CHMPFL13315.POREA 0 EUR
CHMPFL13315.POR
4-Fluorbenzaldehydoxim
4-Fluorbenzaldehydoxim
Formel: C₇H₆FNO
Molecular Weight: 139,13 g/mol
MDL: MFCD00133101
CAS-Nummer: 459-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR