3'-Fluor-4'-hydroxyacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(3-Fluoro-4-hydroxyphenyl)-1-ethanone

FL13316.POR
CHMPFL13316.POREA 0 EUR
CHMPFL13316.POR
3'-Fluor-4'-hydroxyacetophenon
3'-Fluor-4'-hydroxyacetophenon
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00215840
CAS-Nummer: 403-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR