4-Fluor-2-iodanilin

Lieferant: CHEMPUR

FL13319.POR
CHMPFL13319.POREA 0 EUR
CHMPFL13319.POR
4-Fluor-2-iodanilin
4-Fluor-2-iodanilin
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
MDL: MFCD00042584
CAS-Nummer: 61272-76-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR