3-Fluor-p-anisaldehyd

Lieferant: CHEMPUR

Synonyme: 3-Fluor-4-methoxybenzaldehyd, 3-Fluor-4-anisaldehyd

FL13322.POR
CHMPFL13322.POREA 0 EUR
CHMPFL13322.POR
3-Fluor-p-anisaldehyd
3-Fluor-p-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00003349
CAS-Nummer: 351-54-2
EINECS: 206-514-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR