2-Fluor-4-methoxybenzoesäure

Lieferant: CHEMPUR

FL13326.POR
CHMPFL13326.POREA 0 EUR
CHMPFL13326.POR
2-Fluor-4-methoxybenzoesäure
2-Fluoro-4-methoxybenzoic acid
Formel: C₈H₇FO₃
MDL: MFCD00509373
CAS-Nummer: 394-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR