5-Fluor-2-methoxybenzoesäure

Lieferant: CHEMPUR

CFF1126.1 FL13329.POR
CHMPCFF1126.1EA 522 EUR
CHMPCFF1126.1 CHMPFL13329.POR
5-Fluor-2-methoxybenzoesäure
5-Fluor-2-methoxybenzoesäure
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
MDL: MFCD00671765
CAS-Nummer: 394-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR