4-Fluor-2-methoxybenzonitril

Lieferant: CHEMPUR

FL13332.POR 013332-5G 013332-1G 013332-25G
CHMPFL13332.POREA 0 EUR
CHMPFL13332.POR FLUO013332-5G FLUO013332-1G FLUO013332-25G
4-Fluor-2-methoxybenzonitril
4-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
MDL: MFCD04116335
CAS-Nummer: 191014-55-8
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR