5-Fluor-2-methoxybenzonitril

Lieferant: CHEMPUR

FL13333.POR 013333-1G 013333-25G 013333-5G
CHMPFL13333.POREA 0 EUR
CHMPFL13333.POR FLUO013333-1G FLUO013333-25G FLUO013333-5G
5-Fluor-2-methoxybenzonitril
5-Fluor-2-methoxybenzonitril
Formel: C₈H₆FNO
MDL: MFCD02683503
CAS-Nummer: 189628-38-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR