2-Fluor-4-methoxybenzylalkohol

Lieferant: CHEMPUR

FL13334.POR 013334-1G
CHMPFL13334.POREA 0 EUR
CHMPFL13334.POR FLUO013334-1G
2-Fluor-4-methoxybenzylalkohol
2-Fluor-4-methoxybenzylalkohol
Formel: C₈H₉FO₂
MDL: MFCD04116336
CAS-Nummer: 405-09-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR