2-Fluor-5-methoxybenzylalkohol

Lieferant: CHEMPUR

Synonyme: (2-Fluoro-5-methoxyphenyl)methanol

FL13335.POR
CHMPFL13335.POREA 0 EUR
CHMPFL13335.POR
2-Fluor-5-methoxybenzylalkohol
2-Fluor-5-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD04116337
CAS-Nummer: 161643-29-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR