4-Fluor-2-methoxybenzylalkohol

Lieferant: CHEMPUR

Synonyme: (4-Fluoro-2-methoxyphenyl)methanol

FL13336.POR
CHMPFL13336.POREA 0 EUR
CHMPFL13336.POR
4-Fluor-2-methoxybenzylalkohol
4-Fluor-2-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD04116338
CAS-Nummer: 157068-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR