5-Fluor-2-methoxybenzylalkohol

Lieferant: CHEMPUR

Synonyme: (5-Fluoro-2-methoxyphenyl)methanol

FL13338.POR
CHMPFL13338.POREA 0 EUR
CHMPFL13338.POR
5-Fluor-2-methoxybenzylalkohol
5-Fluor-2-methoxybenzylalkohol
Formel: C₈H₉FO₂
Molecular Weight: 156,16 g/mol
MDL: MFCD00671767
CAS-Nummer: 426831-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR