2-Fluor-5-methoxybenzylamin

Lieferant: CHEMPUR

FL13340.POR
CHMPFL13340.POREA 0 EUR
CHMPFL13340.POR
2-Fluor-5-methoxybenzylamin
2-Fluor-5-methoxybenzylamin
Formel: C₈H₁₀FNO
MDL: MFCD04116359
CAS-Nummer: 93071-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR