4-Fluor-2-methoxybenzylamin

Lieferant: CHEMPUR

Synonyme: (4-Fluoro-2-methoxyphenyl)methanamine, (4-Fluoro- 2-methoxy-phenyl) methanamine

FL13342.POR
CHMPFL13342.POREA 0 EUR
CHMPFL13342.POR
4-Fluor-2-methoxybenzylamin
4-Fluor-2-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
MDL: MFCD04116361
CAS-Nummer: 870563-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR