4-Fluor-3-methoxybenzylamin

Lieferant: CHEMPUR

Synonyme: 5-Aminomethyl-2-fluoroanisole, 5-(Aminomethyl)-2-fluoroanisole

FL13343.POR
CHMPFL13343.POREA 0 EUR
CHMPFL13343.POR
4-Fluor-3-methoxybenzylamin
4-Fluor-3-methoxybenzylamin
Formel: C₈H₁₀FNO
Molecular Weight: 155,1700 g/mol
MDL: MFCD04116362
CAS-Nummer: 508177-67-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR