2-Fluor-4-methoxyphenol
Lieferant: CHEMPUR
Synonyme:
3-Fluor-4-hydroxyanisol, 1-Fluor-2-hydroxy-5-methoxybenzol
FL13350.POR
CHMPFL13350.POREA
0
EUR
CHMPFL13350.POR
2-Fluor-4-methoxyphenol
2-Fluor-4-methoxyphenol
Formel:
C₇H₇FO₂ Molecular Weight: 142,13 g/mol |
MDL:
MFCD00070796 CAS-Nummer: 167683-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...