3-Fluor-4-methylanisol

Lieferant: CHEMPUR

Synonyme: 2-Fluoro-4-methoxy-1-methylbenzene

FL13356.POR
CHMPFL13356.POREA 0 EUR
CHMPFL13356.POR
3-Fluor-4-methylanisol
3-Fluor-4-methylanisol
Formel: C₈H₉FO
Molecular Weight: 140,1600 g/mol
MDL: MFCD00143163
CAS-Nummer: 405-06-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR