4-Fluor-2-methylanisol

Lieferant: CHEMPUR
Achtung

Synonyme: 1-Fluor-4-methoxy-3-methylbenzol, 5-Fluor-2-methoxytoluol

FL13357.POR
CHMPFL13357.POREA 0 EUR
CHMPFL13357.POR
4-Fluor-2-methylanisol
4-Fluor-2-methylanisol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
MDL: MFCD00143319
CAS-Nummer: 399-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR