5-Fluor-2-methylanisol

Lieferant: CHEMPUR

FL13358.POR
CHMPFL13358.POREA 0 EUR
CHMPFL13358.POR
5-Fluor-2-methylanisol
5-Fluor-2-methylanisol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
MDL: MFCD00673008
CAS-Nummer: 95729-22-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR