4-Fluor-2-nitroanisol

Lieferant: CHEMPUR
Achtung

Synonyme: 4-Fluoro-1-methoxy-2-nitrobenzene

FL13365.POR
CHMPFL13365.POREA 0 EUR
CHMPFL13365.POR
4-Fluor-2-nitroanisol
4-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013375
CAS-Nummer: 445-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR