2'-Fluor-4'-nonyloxyacetophenon

Lieferant: CHEMPUR

FL13366.POR
CHMPFL13366.POREA 0 EUR
CHMPFL13366.POR
2'-Fluor-4'-nonyloxyacetophenon
2'-Fluor-4'-nonyloxyacetophenon
Formel: C₁₇H₂₅FO₂
MDL: MFCD00142715
CAS-Nummer: 203066-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR