1-Fluor-2,3,5-tribrombenzol

Lieferant: CHEMPUR

Synonyme: 1,2,5-Tribromo-3-fluorobenzene

FL13382.POR
CHMPFL13382.POREA 0 EUR
CHMPFL13382.POR
1-Fluor-2,3,5-tribrombenzol
1-Fluoro-2,3,5-tribromobenzene
Formel: C₆H₂Br₃F
Molecular Weight: 332,79 g/mol
MDL: MFCD00142627
CAS-Nummer: 2839-37-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR