1,2,3-Tribrom-5-fluorbenzol

Lieferant: CHEMPUR
Achtung

FL13383.POR
CHMPFL13383.POREA 0 EUR
CHMPFL13383.POR
1,2,3-Tribrom-5-fluorbenzol
1,2,3-Tribrom-5-fluorbenzol
Formel: C₆H₂Br₃F
Molecular Weight: 332,79 g/mol
MDL: MFCD00061117
CAS-Nummer: 576-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR