6-Fluorveratrumaldehyd

Lieferant: CHEMPUR
Achtung

Synonyme: 3,4-Dimethoxy-6-fluorbenzaldehyd

FL13384.POR
CHMPFL13384.POREA 0 EUR
CHMPFL13384.POR
6-Fluorveratrumaldehyd
6-Fluorveratrumaldehyd
Formel: C₉H₉FO₃
Molecular Weight: 184.17 g/mol
MDL: MFCD00061108
CAS-Nummer: 71924-62-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR