2-(Trifluormethyl)anisol

Lieferant: CHEMPUR
Gefahr

FL13385.POR
CHMPFL13385.POREA 0 EUR
CHMPFL13385.POR
2-(Trifluormethyl)anisol
2-(Trifluormethyl)anisol
Formel: C₈H₇F₃O
Molecular Weight: 176.14 g/mol
MDL: MFCD00142938
CAS-Nummer: 395-48-2
UN: 1993
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR