3-Acetylthiophen
Lieferant: CHEMPUR
Synonyme:
1-(3-Thienyl)-1-ethanon, Methyl-3-thienylketon
FL13392.POR
CHMPFL13392.POREA
0
EUR
CHMPFL13392.POR
3-Acetylthiophen
3-Acetylthiophen
Formel:
C₆H₆OS Molecular Weight: 126,18 g/mol Siedepunkt: 208 °C (1013 hPa) Schmelzpunkt: 56 °C |
MDL:
MFCD00005468 CAS-Nummer: 1468-83-3 EINECS: 215-996-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...