2-Amino-4-chlorbenzothiazol

Lieferant: CHEMPUR

FL13399.POR
CHMPFL13399.POREA 0 EUR
CHMPFL13399.POR
2-Amino-4-chlorbenzothiazol
2-Amino-4-chlorobenzothiazole
Formel: C₇H₅ClN₂S
Molecular Weight: 184,65 g/mol
MDL: MFCD00005791
CAS-Nummer: 19952-47-7
EINECS: 243-439-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR