4-Amino-2-mercaptopyrimidin

Lieferant: CHEMPUR

Synonyme: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine

FL13405.POR
CHMPFL13405.POREA 0 EUR
CHMPFL13405.POR
4-Amino-2-mercaptopyrimidin
4-Amino-2-mercaptopyrimidine
Formel: C₄H₅N₃S
Molecular Weight: 127.1676 g/mol
MDL: MFCD00006075
CAS-Nummer: 333-49-3
EINECS: 206-374-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR