4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO)

Lieferant: CHEMPUR

Synonyme: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine

FL13419.POR
CHMPFL13419.POREA 0 EUR
CHMPFL13419.POR
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO)
4-Amino-2,2,6,6-tetramethylpiperidin-1-oxyl (4-Amino-TEMPO)
Formel: C₉H₁₉N₂O
Molecular Weight: 171,26 g/mol
MDL: MFCD00006479
CAS-Nummer: 14691-88-4
EINECS: 238-738-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR