2,1-Benzisoxazol

Lieferant: CHEMPUR

Synonyme: Anthranil, Benzo[c]isoxazole, 2,1-Benzisoxazole

FL13425.POR
CHMPFL13425.POREA 0 EUR
CHMPFL13425.POR
2,1-Benzisoxazol
2,1-Benzisoxazole
Formel: C₇H₅NO
Molecular Weight: 119,1207 g/mol
MDL: MFCD00005806
CAS-Nummer: 271-58-9
EINECS: 205-980-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR