trans-4-tert-Butylcyclohexancarbonsäure

Lieferant: CHEMPUR

Synonyme: trans-4-(tert-Butyl)cyclohexanecarboxylic acid

FL13464.POR
CHMPFL13464.POREA 0 EUR
CHMPFL13464.POR
trans-4-tert-Butylcyclohexancarbonsäure
trans-4-tert-Butylcyclohexancarbonsäure
Formel: C₁₁H₂₀O₂
Molecular Weight: 184,28 g/mol
MDL: MFCD00466237
CAS-Nummer: 943-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR