5-tert-Butyl-2,4-dihydroxybenzophenon

Lieferant: CHEMPUR

FL13465.POR
CHMPFL13465.POREA 0 EUR
CHMPFL13465.POR
5-tert-Butyl-2,4-dihydroxybenzophenon
5-tert-Butyl-2,4-dihydroxybenzophenon
Formel: C₁₇H₁₈O₃
MDL: MFCD01861910
CAS-Nummer: 4211-67-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR