(4-Chlorphenyl)methansulfonsäurechlorid
Lieferant: CHEMPUR
Synonyme:
4-Chloro-α-toluenesulphonyl chloride
FL13482.POR
020874-1G
020874-5G
CHMPFL13482.POREA
0
EUR
CHMPFL13482.POR
FLUO020874-1G
FLUO020874-5G
(4-Chlorphenyl)methansulfonsäurechlorid
(4-Chlorophenyl)methanesulphonyl chloride
Formel:
C₇H₆Cl₂O₂S Molecular Weight: 225.10 g/mol |
MDL:
MFCD01631927 CAS-Nummer: 6966-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...