Methyl-2,3-diaminobenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 2,3-diaminobenzoate

FL13500.POR
CHMPFL13500.POREA 0 EUR
CHMPFL13500.POR
Methyl-2,3-diaminobenzoat
Methyl-2,3-diaminobenzoat
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,1800 g/mol
MDL: MFCD04038589
CAS-Nummer: 107582-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR