2,6-Dimethoxybenzaldehyd
Lieferant: CHEMPUR
|
Achtung
|
FL13536.POR
CHMPFL13536.POREA
0
EUR
CHMPFL13536.POR
2,6-Dimethoxybenzaldehyd
2,6-Dimethoxybenzaldehyde
Formel:
C₉H₁₀O₃ Molecular Weight: 166.18 g/mol Siedepunkt: 285 Schmelzpunkt: 95…96 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00010862 CAS-Nummer: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...