2,4-Dimethoxybenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 2,4-Dimethoxybenzolsulfochlorid, 2,4-Dimethoxybenzolsulfonsäurechlorid, 2,4-Dimethoxybenzol-1-sulfonylchlorid

FL13537.POR
CHMPFL13537.POREA 0 EUR
CHMPFL13537.POR
2,4-Dimethoxybenzolsulfonylchlorid
2,4-Dimethoxybenzolsulfonylchlorid
Formel: C₈H₉ClO₄S
Molecular Weight: 236,68 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00060683
CAS-Nummer: 63624-28-2
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR