2,3-Dimethylbenzofuran

Lieferant: CHEMPUR

Synonyme: 2,3-Dimethylbenzo[b]furan

FL13541.POR
CHMPFL13541.POREA 0 EUR
CHMPFL13541.POR
2,3-Dimethylbenzofuran
2,3-Dimethylbenzofuran
Formel: C₁₀H₁₀O
Molecular Weight: 146,19 g/mol
MDL: MFCD00078708
CAS-Nummer: 3782-00-1
EINECS: 223-245-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR