1-Ethoxynaphthalin
Lieferant: CHEMPUR
Synonyme:
Ethyl-1-naphthylether
FL13566.POR
CHMPFL13566.POREA
0
EUR
CHMPFL13566.POR
1-Ethoxynaphthalin
1-Ethoxynaphthalin
Formel:
C₁₂H₁₂O Molecular Weight: 172,23 g/mol Siedepunkt: 280 °C (1013 hPa) Schmelzpunkt: 5,5 °C Dichte: 1,0711 g/cm³ (4 °C) |
MDL:
MFCD00003928 CAS-Nummer: 5328-01-8 EINECS: 226-213-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...