Benzothiazol-2-ylmethanol

Lieferant: CHEMPUR

Synonyme: 2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole

FL13600.POR
CHMPFL13600.POREA 0 EUR
CHMPFL13600.POR
Benzothiazol-2-ylmethanol
Benzothiazol-2-ylmethanol
Formel: C₈H₇NOS
Molecular Weight: 165,21 g/mol
MDL: MFCD00226293
CAS-Nummer: 37859-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR