6-Hydroxy-2-naphthoesäure

Lieferant: CHEMPUR

Synonyme: 6-hydroxynaphthalene-2-carboxylic acid

FL13602.POR
CHMPFL13602.POREA 0 EUR
CHMPFL13602.POR
6-Hydroxy-2-naphthoesäure
6-Hydroxy-2-naphthoesäure
Formel: C₁₁H₈O₃
Molecular Weight: 188,18 g/mol
MDL: MFCD00060070
CAS-Nummer: 16712-64-4
EINECS: 240-759-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR