5-Nitrosalicylaldehyd
Lieferant: CHEMPUR
|
Achtung
|
Synonyme:
2-Hydroxy-5-nitrobenzaldehyde
FL13603.POR
CHMPFL13603.POREA
0
EUR
CHMPFL13603.POR
5-Nitrosalicylaldehyd
5-Nitrosalicylaldehyd
Formel:
C₇H₅NO₄ Molecular Weight: 167,12 g/mol Schmelzpunkt: 123…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007337 CAS-Nummer: 97-51-8 EINECS: 202-587-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...