4-Chinazolon

Lieferant: CHEMPUR

Synonyme: 4-Hydroxyquinazoline, Quinazolin-4(3H)-one

FL13606.POR
CHMPFL13606.POREA 0 EUR
CHMPFL13606.POR
4-Chinazolon
4-Chinazolon
Formel: C₈H₆N₂O
Molecular Weight: 146,15 g/mol
MDL: MFCD00511302
CAS-Nummer: 491-36-1
EINECS: 207-735-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR